Assay Details
Target Gene Details
Entrez Gene ID: | 320712 |
Gene Name: | ABI gene family, member 3 (NESH) binding protein |
Gene Aliases: |
5033411B22Rik, AI506287, BG172926, D930038M13Rik, TARSH, eratin |
Location: |
Chr.16:56477801-56690135 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abi3bp | NM_001014399.2 | 2 | 202 | NP_001014421.2 |
| NM_001014422.2 | 2 | 202 | NP_001014422.1 | |
| NM_001014423.2 | 2 | 202 | NP_001014423.1 | |
| NM_001014424.2 | 2 | 202 | NP_001014424.1 | |
| NM_178790.4 | 2 | 202 | NP_848905.2 | |
| AB190339.1 | 2 | 148 | BAD91388.1 | |
| AB190340.1 | 2 | 148 | BAD91389.1 | |
| AB190341.1 | 2 | 148 | BAD91390.1 | |
| AB190342.1 | 2 | 148 | BAD91391.1 | |
| AB190343.1 | 2 | 148 | BAD91392.1 | |
| AK041192.1 | 2 | 198 | BAC30856.2 | |
| AK043879.1 | 2 | 176 | ||
| AK051214.1 | 2 | 172 | BAC34560.1 | |
| AK086582.1 | 2 | 205 | BAC39696.1 | |
| BB663305.1 | 2 | 202 | ||
| BC145388.1 | 2 | 165 | ||
| BC150763.1 | 2 | 165 |

Genomic Map