Assay Details
Target Gene Details
Entrez Gene ID: | 12053 |
Gene Name: | B cell leukemia/lymphoma 6 |
Gene Aliases: |
Bcl5 |
Location: |
Chr.16:23965052-23988612 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl6 | NM_009744.3 | 7 | 1668 | NP_033874.1 |
| AK036975.1 | 7 | 1692 | BAC29654.1 | |
| AK039228.1 | 7 | 1690 | BAC30286.1 | |
| AK151229.1 | 7 | 1693 | BAE30221.1 | |
| BC052315.1 | 7 | 1669 | AAH52315.1 | |
| D38377.1 | 7 | 1657 | BAA07456.1 | |
| U41465.1 | 6 | 1680 | AAB17432.1 |
More Information
Set Membership: |
|

Genomic Map