Assay Details
Target Gene Details
Entrez Gene ID: | 17250 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 1 |
Gene Aliases: |
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1 |
Location: |
Chr.16:14361521-14474879 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcc1 | NM_008576.3 | NP_032602.1 | ||
| AF022908.1 | AAB80938.1 | |||
| AK029876.1 | BAC26654.1 | |||
| AK150015.1 | BAE29240.1 | |||
| AK152781.1 | ||||
| BC090617.1 |
More Information
Set Membership: |
|

Genomic Map