Assay Details
Target Gene Details
Entrez Gene ID: | 17250 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 1 |
Gene Aliases: |
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1 |
Location: |
Chr.16:14361521-14474879 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcc1 | NM_008576.3 | 24 | 3519 | NP_032602.1 |
| AF022908.1 | 24 | 3481 | AAB80938.1 | |
| AK029876.1 | 24 | 3520 | BAC26654.1 | |
| AK033750.1 | 2 | 302 | ||
| AK049174.1 | 8 | 1286 | BAC33586.1 | |
| AK140421.1 | 2 | 296 | ||
| BC090617.1 | 24 | 3490 |
More Information
Set Membership: |
|

Genomic Map