Assay Details
Target Gene Details
Entrez Gene ID: | 16423 |
Gene Name: | CD47 antigen (Rh-related antigen, integrin-associated signal transducer) |
Gene Aliases: |
9130415E20Rik, AA407862, AI848868, AW108519, B430305P08Rik, IAP, Itgp |
Location: |
Chr.16:49855252-49912424 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd47 | NM_010581.3 | 7 | 927 | NP_034711.1 |
| AB012693.1 | 7 | 866 | BAA25401.1 | |
| AK151922.1 | 6 | 810 | BAE30800.1 | |
| BC009094.1 | 5 | 493 | AAH09094.1 | |
| BC012667.1 | 7 | 861 | AAH12667.1 | |
| HQ585874.1 | 6 | 731 | ||
| Z25524.1 | 7 | 927 | CAA80978.1 |
More Information
Set Membership: |
|

Genomic Map