Assay Details
Target Gene Details
Entrez Gene ID: | 17250 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 1 |
Gene Aliases: |
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1 |
Location: |
Chr.16:14361521-14474879 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcc1 | NM_008576.3 | 8 | 877 | NP_032602.1 |
| AF022908.1 | 8 | 839 | AAB80938.1 | |
| AK029876.1 | 8 | 878 | BAC26654.1 | |
| AK150015.1 | 8 | 858 | BAE29240.1 | |
| AK152781.1 | 8 | 858 | ||
| AK162656.1 | 2 | 2816 | ||
| BC090617.1 | 8 | 848 |
More Information
Set Membership: |
|

Genomic Map