Assay Details
Target Gene Details
Entrez Gene ID: | 27416 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 5 |
Gene Aliases: |
2900011L11Rik, AI132311, Abcc5a, Abcc5b, Mrp5 |
Location: |
Chr.16:20331303-20426394 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcc5 | NM_013790.2 | NP_038818.2 | ||
| AB019003.1 | BAA76609.1 | |||
| BC090629.1 | AAH90629.1 |
More Information
Set Membership: |
|

Genomic Map