Assay Details
Target Gene Details
Entrez Gene ID: | 100126824 |
Gene Name: | SCO cytochrome oxidase deficient homolog 2 (yeast) |
Gene Aliases: |
- |
Location: |
Chr.15:89371637-89373818 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sco2 | NM_001111288.1 | NP_001104758.1 | ||
| AK013765.1 | BAB28986.1 | |||
| AK186876.1 | ||||
| BC019554.1 | AAH19554.1 |
Target Gene Details
Entrez Gene ID: | 72962 |
Gene Name: | thymidine phosphorylase |
Gene Aliases: |
2900072D10Rik, Ecgf1, PD-ECGF, PDECGF, Pdgfec |
Location: |
Chr.15:89371931-89377036 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tymp | NM_138302.1 | NP_612175.1 | ||
| AB060274.1 | BAB41208.1 |
More Information
Set Membership: |
|

Genomic Map