Assay Details
Target Gene Details
Entrez Gene ID: | 72962 |
Gene Name: | thymidine phosphorylase |
Gene Aliases: |
2900072D10Rik, Ecgf1, PD-ECGF, PDECGF, Pdgfec |
Location: |
Chr.15:89371931-89377036 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tymp | NM_138302.1 | 5 | 607 | NP_612175.1 |
| AB060274.1 | 5 | 607 | BAB41208.1 |
More Information
Set Membership: |
|

Genomic Map