Assay Details
Target Gene Details
Entrez Gene ID: | 223669 |
Gene Name: | zinc finger protein 7 |
Gene Aliases: |
KRAB20, KRAB7, Krox-2, Zfp-7, Zfp65, Zfp80, Zfp86-rs1, mszf73-2 |
Location: |
Chr.15:76879217-76892395 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp7 | NM_145916.2 | NP_666021.2 | ||
| AK038299.1 | 1 | 1528 | ||
| AK143414.1 | ||||
| AK144887.1 | BAE26117.1 | |||
| AK168948.1 | BAE40754.1 | |||
| BC011501.1 | AAH11501.1 | |||
| BU613201.1 |
More Information
Set Membership: |
|

Genomic Map