Assay Details
Target Gene Details
| Entrez Gene ID: | 100038602 | 
| Gene Name: | predicted gene 10865 | 
| Gene Aliases: | ENSMUSG00000075558 | 
| Location: | Chr.15:78994955-79011041 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Gm10865 | NR_045746.1 | |||
| AK139534.1 | BAE24054.1 | 
Target Gene Details
| Entrez Gene ID: | 110253 | 
| Gene Name: | TRIO and F-actin binding protein | 
| Gene Aliases: | AI428493, EST478828, Tara, mKIAA1662 | 
| Location: | Chr.15:78947724-79005869 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 23 - Exon 24 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Triobp | NM_001024716.1 | NP_001019887.1 | ||
| NM_001039156.1 | NP_001034245.1 | |||
| NM_138579.4 | NP_613045.3 | |||
| AK020187.1 | ||||
| AK053194.1 | BAC35308.1 | |||
| AK085512.1 | ||||
| AK129417.1 | BAC98227.2 | |||
| AK170390.1 | BAE41764.1 | |||
| BC003984.1 | AAH03984.1 | |||
| BC042760.1 | AAH42760.1 | |||
| BC053329.1 | AAH53329.1 | |||
| BU058377.1 | ||||
| CA322031.1 | ||||
| DQ228000.1 | ABB59556.1 | |||
| DQ228001.2 | ABB59557.2 | |||
| DQ278602.1 | ABB77203.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top