Assay Details
Target Gene Details
Entrez Gene ID: | 12856 |
Gene Name: | cytochrome c oxidase assembly protein 17 |
Gene Aliases: |
AI037035 |
Location: |
Chr.16:38346984-38352763 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox17 | NM_001017429.2 | NP_001017429.1 | ||
| AK146388.1 | BAE27132.1 | |||
| AK148493.1 | BAE28585.1 | |||
| AK152968.1 | BAE31628.1 | |||
| AK158051.1 | BAE34337.1 | |||
| AK160687.1 | 1 | 2157 | ||
| BC048668.1 | AAH48668.1 | |||
| BG792513.1 |
More Information
Set Membership: |
|

Genomic Map