Assay Details
Target Gene Details
Entrez Gene ID: | 74185 |
Gene Name: | glucan (1,4-alpha-), branching enzyme 1 |
Gene Aliases: |
2310045H19Rik, 2810426P10Rik, D16Ertd536e |
Location: |
Chr.16:70313949-70569720 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbe1 | NM_028803.4 | 8 | 1254 | NP_083079.1 |
| AK009815.1 | 8 | 1070 | BAB26519.1 | |
| AK050365.1 | 8 | 1141 | BAC34210.1 | |
| AK050423.1 | 8 | 1140 | BAC34248.1 | |
| AK148186.1 | 2 | 182 | ||
| BC017541.1 | 8 | 1115 | AAH17541.1 |
More Information
Set Membership: |
|

Genomic Map