Assay Details
Target Gene Details
Entrez Gene ID: | 14811 |
Gene Name: | glutamate receptor, ionotropic, NMDA2A (epsilon 1) |
Gene Aliases: |
GluN2A, NMDAR2A, NR2A |
Location: |
Chr.16:9573737-9996500 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grin2a | NM_008170.2 | 5 | 1372 | NP_032196.2 |
| D10217.2 | 5 | 1372 | BAA01069.2 |
More Information
Set Membership: |
|

Genomic Map