Assay Details
Target Gene Details
Entrez Gene ID: | 72599 |
Gene Name: | protein disulfide isomerase associated 5 |
Gene Aliases: |
2700053F16Rik, AU015525, Pdir |
Location: |
Chr.16:35397308-35490898 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdia5 | NM_028295.1 | NP_082571.1 | ||
| BC009151.1 | AAH09151.1 |
More Information
Set Membership: |
|

Genomic Map