Assay Details
Target Gene Details
Entrez Gene ID: | 68172 |
Gene Name: | ribosomal protein L39-like |
Gene Aliases: |
4930517K11Rik |
Location: |
Chr.16:10170192-10174911 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl39l | NM_026594.2 | NP_080870.1 | ||
| AK005645.1 | BAB24165.1 | |||
| AK019681.1 | BAB31834.1 | |||
| BC023885.1 | ||||
| BC051469.1 | AAH51469.1 |
More Information
Set Membership: |
|

Genomic Map