Assay Details
Target Gene Details
Entrez Gene ID: | 106298 |
Gene Name: | RRN3 RNA polymerase I transcription factor homolog (yeast) |
Gene Aliases: |
AL023001, E130302O19Rik, R75565, TIF-1A, Tif1a |
Location: |
Chr.16:13780699-13814841 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrn3 | NM_001039521.1 | NP_001034610.1 | ||
| AK080397.1 | ||||
| AK165676.1 | ||||
| AK211678.1 | ||||
| BC034110.1 | AAH34110.1 | |||
| BC138774.1 | ||||
| BC138776.1 |
More Information
Set Membership: |
|

Genomic Map