Assay Details
Target Gene Details
Entrez Gene ID: | 56637 |
Gene Name: | glycogen synthase kinase 3 beta |
Gene Aliases: |
7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3 |
Location: |
Chr.16:38089001-38246080 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsk3b | NM_019827.6 | 5 | 2064 | NP_062801.1 |
| AB066114.1 | 6 | 2063 | BAD86827.1 | |
| AF156099.2 | 5 | 705 | AAD39258.2 | |
| AK154293.1 | 6 | 1542 | BAE32494.1 | |
| AK164048.1 | 4 | 416 | ||
| AK170965.1 | 6 | 1511 | BAE42146.1 | |
| BC006936.1 | 5 | 571 | AAH06936.1 | |
| BC060743.1 | 6 | 1991 | AAH60743.1 |
More Information
Set Membership: |
|

Genomic Map