Assay Details
Target Gene Details
Entrez Gene ID: | 66409 |
Gene Name: | ribosomal L1 domain containing 1 |
Gene Aliases: |
2410005K20Rik, C76439, pBK1 |
Location: |
Chr.16:11193037-11203292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsl1d1 | NM_025546.2 | NP_079822.1 | ||
| AK012937.1 | BAB28559.1 | |||
| AK075933.1 | BAC36063.1 | |||
| AK136025.1 | BAE22779.1 | |||
| AK145699.1 | BAE26596.1 | |||
| AK162241.1 | BAE36811.1 | |||
| AK165572.1 | BAE38264.1 | |||
| AK166479.1 | BAE38799.1 | |||
| AK166579.1 | BAE38868.1 | |||
| AK168469.1 | BAE40361.1 | |||
| AK168717.1 | BAE40559.1 | |||
| AK171805.1 | BAE42673.1 | |||
| BC034355.1 | AAH34355.1 | |||
| DQ445383.1 | ABE03015.1 |
More Information
Set Membership: |
|

Genomic Map