Assay Details
Target Gene Details
Entrez Gene ID: | 66053 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 2 |
Gene Aliases: |
0610009L05Rik, 1700016N17Rik, 4921520K19Rik, 4930511F14Rik, AA589416, C130078A06Rik, CYC4 |
Location: |
Chr.16:17086555-17111263 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppil2 | NM_001252444.1 | NP_001239373.1 | ||
| NM_001252445.1 | NP_001239374.1 | |||
| NM_144954.3 | NP_659203.1 | |||
| AK009460.1 | BAB26301.1 | |||
| AK012472.1 | ||||
| AK081792.1 | BAC38333.1 | |||
| AK090052.1 | BAC41068.1 | |||
| AK161596.1 | BAE36483.1 | |||
| BC028899.1 | AAH28899.1 |
More Information
Set Membership: |
|

Genomic Map