Assay Details
Target Gene Details
Entrez Gene ID: | 70757 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 2 |
Gene Aliases: |
6330408J20Rik, AI255777, AI481689, Hcad2, Ptplb |
Location: |
Chr.16:35022421-35109190 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hacd2 | NM_023587.2 | NP_076076.2 | ||
| AF169286.1 | AAF29489.1 | |||
| AK018144.1 | BAB31092.1 | |||
| AK148143.1 | BAE28371.1 | |||
| BC027289.1 | AAH27289.1 | |||
| BC131899.1 | ||||
| BC131901.1 |

Genomic Map