Assay Details
Target Gene Details
Entrez Gene ID: | 72058 |
Gene Name: | immunoglobulin superfamily, member 5 |
Gene Aliases: |
2010003D20Rik, JCAM, Jam4 |
Location: |
Chr.16:96360467-96465572 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf5 |
Target Gene Details
Entrez Gene ID: | 16415 |
Gene Name: | integrin beta 2-like |
Gene Aliases: |
5033406G21Rik, pactolus |
Location: |
Chr.16:96422295-96444365 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb2l | NM_008405.3 | NP_032431.2 | ||
| AF051367.1 | AAC25502.1 | |||
| AK036640.1 | ||||
| AK137534.1 | BAE23399.1 | |||
| AK158108.1 | ||||
| BC113142.1 | AAI13143.1 |
More Information
Set Membership: |
|

Genomic Map