Assay Details
Target Gene Details
Entrez Gene ID: | 107589 |
Gene Name: | myosin, light polypeptide kinase |
Gene Aliases: |
9530072E15Rik, A930019C19Rik, AW489456, KRP, MLCK108, MLCK210, Mlck, nmMlck, smMLCK |
Location: |
Chr.16:34745199-35002436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mylk | NM_139300.3 | NP_647461.3 | ||
| AY237726.1 | AAO85807.1 | |||
| BC058610.1 | AAH58610.2 | |||
| BC145279.1 | ||||
| BC150749.1 |

Genomic Map