Assay Details
Target Gene Details
Entrez Gene ID: | 12394 |
Gene Name: | runt related transcription factor 1 |
Gene Aliases: |
AML1, CBF-alpha-2, Cbfa2, Pebp2a2, Pebpa2b |
Location: |
Chr.16:92601466-92826074 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Runx1 | NM_001111021.2 | 7 | 1092 | NP_001104491.1 |
| NM_001111022.2 | 6 | 900 | NP_001104492.1 | |
| NM_001111023.2 | 5 | 2427 | NP_001104493.1 | |
| NM_009821.3 | 4 | 2235 | NP_033951.2 | |
| AK051758.1 | 4 | 1773 | BAC34757.1 | |
| AK137493.1 | 7 | 1062 | ||
| AK145098.1 | 7 | 1085 | BAE26233.1 | |
| AK165307.1 | 7 | 1083 | ||
| BC069929.1 | 6 | 837 | AAH69929.1 | |
| D13802.1 | 5 | 1105 | BAA02960.1 | |
| D26532.1 | 4 | 2232 | BAA05535.1 |
More Information
Set Membership: |
|

Genomic Map