Assay Details
Target Gene Details
Entrez Gene ID: | 93734 |
Gene Name: | Mpv17 transgene, kidney disease mutant-like |
Gene Aliases: |
M-LP |
Location: |
Chr.16:13903161-13949619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpv17l | NM_001289561.1 | NP_001276490.1 | ||
| NM_001289562.1 | NP_001276491.1 | |||
| NM_001289563.1 | NP_001276492.1 | |||
| NM_001289565.1 | NP_001276494.1 | |||
| NM_001289567.1 | NP_001276496.1 | |||
| NM_033564.3 | NP_291042.2 | |||
| AF305634.1 | AAK32113.2 | |||
| AK136449.1 | BAE22983.1 | |||
| AK144372.1 | BAE25854.1 | |||
| AY513273.1 | AAS82777.1 | |||
| BB618586.1 | ||||
| BC037713.1 | AAH37713.1 | |||
| BC094450.1 | AAH94450.1 |
More Information
Set Membership: |
|

Genomic Map