Assay Details
Target Gene Details
Entrez Gene ID: | 72117 |
Gene Name: | N(alpha)-acetyltransferase 50, NatE catalytic subunit |
Gene Aliases: |
2600005K24Rik, 2810441M03Rik, AW112078, Mak3, Mak3p, Nat13, Nat5, San |
Location: |
Chr.16:44139809-44163366 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa50 | NM_028108.3 | 2 | 364 | NP_082384.1 |
| AK011160.1 | 2 | 290 | BAB27439.1 | |
| AK135658.1 | 2 | 307 | BAE22602.1 | |
| AK145345.1 | 2 | 292 | BAE26378.1 | |
| AK156791.1 | 2 | 310 | BAE33858.1 | |
| AK167141.1 | 3 | 484 | BAE39286.1 | |
| AK168394.1 | 2 | 335 | BAE40319.1 | |
| AK184762.1 | 1 | 60 | ||
| AK195449.1 | 1 | 60 | ||
| AK198408.1 | 2 | 133 | ||
| AK219650.1 | 1 | 60 | ||
| BC046283.1 | 2 | 284 | AAH46283.1 | |
| BC055846.1 | 2 | 129 | AAH55846.1 | |
| BC057117.1 | 2 | 323 | AAH57117.1 | |
| BY062588.1 |
More Information
Set Membership: |
|

Genomic Map