Assay Details
Target Gene Details
Entrez Gene ID: | 57263 |
Gene Name: | resistin like beta |
Gene Aliases: |
9030012B21Rik, Fizz2, RELMbeta, Relmb, Xcp3 |
Location: |
Chr.16:48816856-48818892 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Retnlb | NM_023881.4 | NP_076370.3 | ||
| AF290871.1 | AAK83103.1 | |||
| AF323083.1 | AAG59826.1 | |||
| AK020240.1 | BAB32036.1 | |||
| BC022650.1 | AAH22650.1 | |||
| DQ157777.1 | AAZ82036.1 |

Genomic Map