Assay Details
Target Gene Details
Entrez Gene ID: | 106344 |
Gene Name: | replication factor C (activator 1) 4 |
Gene Aliases: |
A1, AI894123, AU040575, RFC37 |
Location: |
Chr.16:23113948-23127730 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfc4 | NM_145480.1 | NP_663455.1 | ||
| AK146954.1 | BAE27563.1 | |||
| AK147030.1 | BAE27622.1 | |||
| BC003335.1 | AAH03335.1 |
More Information
Set Membership: |
|

Genomic Map