Assay Details
Target Gene Details
Entrez Gene ID: | 171286 |
Gene Name: | solute carrier family 12 (potassium/chloride transporters), member 8 |
Gene Aliases: |
CCC9, E330020C02Rik |
Location: |
Chr.16:33517333-33664135 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a8 | NM_001083902.1 | 4 | 540 | NP_001077371.1 |
| NM_134251.2 | 4 | 509 | NP_599012.2 | |
| AF319951.1 | 4 | 503 | AAL37178.1 | |
| BC030926.1 | 4 | 540 | AAH30926.1 |
More Information
Set Membership: |
|

Genomic Map