Assay Details
Target Gene Details
Entrez Gene ID: | 100043314 |
Gene Name: | T cell immunoreceptor with Ig and ITIM domains |
Gene Aliases: |
ENSMUSG00000071552, Vstm3 |
Location: |
Chr.16:43648861-43664184 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tigit | NM_001146325.1 | NP_001139797.1 | ||
CV307751.1 | ||||
EU675311.1 |
