Assay Details
Target Gene Details
Entrez Gene ID: | 83396 |
Gene Name: | GLIS family zinc finger 2 |
Gene Aliases: |
Gli5, Klf16, Nkl |
Location: |
Chr.16:4594713-4625141 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glis2 |
Target Gene Details
Entrez Gene ID: | 66449 |
Gene Name: | presequence translocase-asssociated motor 16 homolog (S. cerevisiae) |
Gene Aliases: |
2010110I09Rik, AV006767, CGI-136, Magmas, Tim16, Timm16 |
Location: |
Chr.16:4616466-4624946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pam16 | NM_025571.1 | NP_079847.1 | ||
| AB073618.1 | BAB91135.1 | |||
| AF349454.1 | AAL57766.1 | |||
| AK003227.1 | BAB22656.1 | |||
| AK008380.1 | BAB25635.1 | |||
| BC024346.1 | AAH24346.1 | |||
| BC096419.1 | AAH96419.1 |
More Information
Set Membership: |
|

Genomic Map