Assay Details
Target Gene Details
Entrez Gene ID: | 72084 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class X |
Gene Aliases: |
2010319C14Rik, PIG-X |
Location: |
Chr.16:32084416-32099727 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigx | NM_001111025.1 | NP_001104495.1 | ||
| NM_024464.3 | NP_077784.2 | |||
| AK008583.2 | BAC25223.1 | |||
| BC002202.1 | AAH02202.1 | |||
| CF581933.1 |

Genomic Map