Assay Details
Target Gene Details
Entrez Gene ID: | 13972 |
Gene Name: | guanine nucleotide binding protein (G protein), beta polypeptide 1-like |
Gene Aliases: |
ESTM55, Gm16314, Me49f07, OTTMUSG00000033458, Wdr14, Wdvcf |
Location: |
Chr.16:18498021-18566680 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnb1l | NM_001081682.2 | 5 | 584 | NP_001075151.1 |
| NM_001285491.1 | 5 | 580 | NP_001272420.1 | |
| NM_001285493.1 | 5 | 522 | NP_001272422.1 | |
| NM_001285494.1 | 5 | 526 | NP_001272423.1 | |
| NM_023120.5 | 5 | 526 | NP_075609.2 | |
| NR_104347.1 | 5 | 526 | ||
| AF301595.1 | 5 | 477 | AAG44996.1 | |
| AK010489.1 | 5 | 484 | BAB26980.1 | |
| AK016952.1 | 5 | 510 | ||
| AK087954.1 | 5 | 583 | ||
| AK158210.1 | 5 | 509 | BAE34406.1 | |
| AK166195.1 | 5 | 510 | BAE38622.1 | |
| AK171253.1 | 5 | 506 | BAE42345.1 | |
| AK173227.1 | 5 | 509 | BAD32505.1 | |
| BC024635.1 | 5 | 495 | AAH24635.1 | |
| BC037676.1 | 4 | 412 | AAH37676.1 |
More Information
Set Membership: |
|

Genomic Map