Assay Details
Target Gene Details
Entrez Gene ID: | 13972 |
Gene Name: | guanine nucleotide binding protein (G protein), beta polypeptide 1-like |
Gene Aliases: |
ESTM55, Gm16314, Me49f07, OTTMUSG00000033458, Wdr14, Wdvcf |
Location: |
Chr.16:18498021-18566680 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnb1l | NM_001081682.2 | 7 | 825 | NP_001075151.1 |
| NM_001285491.1 | 7 | 821 | NP_001272420.1 | |
| NM_001285493.1 | 8 | 860 | NP_001272422.1 | |
| NM_001285494.1 | NP_001272423.1 | |||
| NM_023120.5 | 7 | 767 | NP_075609.2 | |
| NR_104347.1 | 8 | 892 | ||
| AF301595.1 | 7 | 718 | AAG44996.1 | |
| AK010489.1 | 7 | 725 | BAB26980.1 | |
| AK016952.1 | 7 | 751 | ||
| AK087954.1 | 7 | 820 | ||
| AK158210.1 | 8 | 847 | BAE34406.1 | |
| AK166195.1 | BAE38622.1 | |||
| AK171253.1 | 7 | 747 | BAE42345.1 | |
| AK173227.1 | 7 | 750 | BAD32505.1 | |
| BC024635.1 | 8 | 861 | AAH24635.1 | |
| BC037676.1 | 6 | 653 | AAH37676.1 |
More Information
Set Membership: |
|

Genomic Map