Assay Details
Target Gene Details
Entrez Gene ID: | 208171 |
Gene Name: | transmembrane serine protease 7 |
Gene Aliases: |
B230219I23Rik, Gm1748 |
Location: |
Chr.16:45656309-45698553 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss7 | NM_172455.3 | 15 | 1833 | NP_766043.3 |
| AK045663.1 | 13 | 1583 | BAC32448.1 | |
| DQ061860.1 | 15 | 1834 | AAY66996.1 |
More Information
Set Membership: |
|

Genomic Map