Assay Details
Target Gene Details
Entrez Gene ID: | 22061 |
Gene Name: | transformation related protein 63 |
Gene Aliases: |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
Location: |
Chr.16:25683763-25892103 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp63 | NM_001127259.1 | NP_001120731.1 | ||
| NM_001127260.1 | NP_001120732.1 | |||
| NM_001127261.1 | NP_001120733.1 | |||
| NM_001127262.1 | NP_001120734.1 | |||
| NM_001127263.1 | NP_001120735.1 | |||
| NM_001127264.1 | NP_001120736.1 | |||
| NM_001127265.1 | NP_001120737.1 | |||
| NM_011641.2 | NP_035771.1 | |||
| AB010152.1 | BAA32432.1 | |||
| AF075434.1 | AAC62639.1 | |||
| AF075435.1 | AAC62640.1 | |||
| AF075436.1 | AAC62641.1 | |||
| AF075437.1 | AAC62642.1 | |||
| AF075438.1 | AAC62643.1 | |||
| AF075439.1 | AAC62644.1 | |||
| AK137253.1 | BAE23286.1 | |||
| AK140023.1 | ||||
| BC090649.1 | AAH90649.1 | |||
| BC092537.1 | AAH92537.1 |
More Information
Set Membership: |
|

Genomic Map