Assay Details
Target Gene Details
Entrez Gene ID: | 319765 |
Gene Name: | insulin-like growth factor 2 mRNA binding protein 2 |
Gene Aliases: |
C330012H03Rik, IMP-2, Imp2, Neilsen |
Location: |
Chr.16:22059009-22163299 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2bp2 | NM_183029.2 | NP_898850.2 | ||
| AK044984.1 | ||||
| AK049196.1 | ||||
| AK077404.1 | ||||
| AK159654.1 | ||||
| AK170531.1 | BAE41861.1 | |||
| AY531659.1 | AAT01428.1 | |||
| CB290059.1 |
More Information
Set Membership: |
|

Genomic Map