Assay Details
Target Gene Details
Entrez Gene ID: | 80859 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta |
Gene Aliases: |
AA408868, INAP, Mail |
Location: |
Chr.16:55811377-55838641 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkbiz | NM_001159395.1 | 1 | 94 | NP_001152867.1 |
| NM_030612.3 | 1 | 94 | NP_085115.1 | |
| AB020974.1 | 1 | 308 | BAB18302.1 | |
| AB047549.2 | 1 | 107 | BAB32782.1 | |
| AK149946.1 | 1 | 90 |
More Information
Set Membership: |
|

Genomic Map