Assay Details
Target Gene Details
Entrez Gene ID: | 80859 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta |
Gene Aliases: |
AA408868, INAP, Mail |
Location: |
Chr.16:55811377-55838641 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkbiz | NM_001159394.1 | NP_001152866.1 | ||
| NM_001159395.1 | NP_001152867.1 | |||
| NM_030612.3 | NP_085115.1 | |||
| AB020974.1 | BAB18302.1 | |||
| AB026551.2 | BAA95161.2 | |||
| AB047549.2 | BAB32782.1 | |||
| AK082908.1 | BAC38681.1 | |||
| AK136509.1 | BAE23017.1 | |||
| AK141318.1 | BAE24648.1 | |||
| AK149946.1 | ||||
| AK150519.1 | BAE29631.1 | |||
| AK152016.1 | BAE30878.1 | |||
| AK156826.1 | ||||
| AK159093.1 | BAE34809.1 | |||
| AK159171.1 | BAE34871.1 | |||
| AK159459.1 | BAE35101.1 | |||
| AK159645.1 | BAE35257.1 | |||
| AK159917.1 | BAE35480.1 | |||
| AK160065.1 | BAE35601.1 | |||
| BC058188.1 | AAH58188.1 |
More Information
Set Membership: |
|

Genomic Map