Assay Details
Target Gene Details
Entrez Gene ID: | 77782 |
Gene Name: | polymerase (DNA directed), theta |
Gene Aliases: |
A430110D14Rik |
Location: |
Chr.16:37011742-37095419 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polq | NM_001159369.1 | 6 | 978 | NP_001152841.1 |
| NM_029977.2 | 11 | 1815 | NP_084253.1 | |
| AK156001.1 | 2 | 283 | BAE33540.1 | |
| AY074936.1 | 11 | 1815 | AAL77225.1 | |
| AY147862.1 | 6 | 978 | AAN64234.1 | |
| AY147863.1 | 12 | 1911 | AAN64235.1 | |
| AY147864.1 | 7 | 1074 | AAN64236.1 | |
| BC138360.1 | 11 | 1756 | ||
| BC157900.1 | 11 | 1756 |
More Information
Set Membership: |
|

Genomic Map