Assay Details
Target Gene Details
Entrez Gene ID: | 73694 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7 |
Gene Aliases: |
2410091C18Rik, AL033374, PRO1853 |
Location: |
Chr.17:78937106-78948052 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf7 | NM_028611.3 | 5 | 531 | NP_082887.2 |
| AK010752.1 | 5 | 530 | BAB27158.2 | |
| AK090097.1 | 5 | 530 | BAC41089.1 | |
| BC108350.1 | 5 | 518 | AAI08351.1 | |
| CX236822.1 | 5 | 531 |

Genomic Map