Assay Details
Target Gene Details
Entrez Gene ID: | 620292 |
Gene Name: | contactin associated protein-like 5C |
Gene Aliases: |
Caspr5-3, EG620292, Gm325 |
Location: |
Chr.17:57769077-58410429 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntnap5c | NM_001081653.1 | NP_001075122.1 |
