Assay Details
Target Gene Details
Entrez Gene ID: | 107971 |
Gene Name: | fibroblast growth factor receptor substrate 3 |
Gene Aliases: |
4930417B13Rik, AI449674, Frs2beta, Snt2 |
Location: |
Chr.17:47687296-47704286 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frs3 | NM_144939.2 | NP_659188.1 | ||
| BC014819.1 | AAH14819.1 |

Genomic Map