Assay Details
Target Gene Details
Entrez Gene ID: | 13177 |
Gene Name: | enoyl-Coenzyme A delta isomerase 1 |
Gene Aliases: |
Dci, eci |
Location: |
Chr.17:24426683-24439316 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eci1 | NM_010023.4 | 6 | 676 | NP_034153.2 |
| AK004838.1 | 6 | 677 | BAB23605.1 | |
| AK029481.1 | 6 | 677 | BAC26469.1 | |
| AK077386.1 | 6 | 667 | ||
| BC022712.1 | 6 | 660 | AAH22712.1 | |
| BC054444.1 | 6 | 663 | AAH54444.1 | |
| Z14049.1 | 6 | 656 | CAA78417.1 |
More Information
Set Membership: |
|

Genomic Map