Assay Details
Target Gene Details
Entrez Gene ID: | 73242 |
Gene Name: | alpha tubulin acetyltransferase 1 |
Gene Aliases: |
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT |
Location: |
Chr.17:35897595-35910093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atat1 | NM_001142744.1 | NP_001136216.1 | ||
| AK011350.1 | ||||
| AK011750.2 | ||||
| AK011838.1 | ||||
| AK028297.1 | BAC25866.1 | |||
| BC028847.1 | AAH28847.1 | |||
| BC030669.1 | AAH30669.1 | |||
| BQ748472.1 |

Genomic Map