Assay Details
Target Gene Details
Entrez Gene ID: | 11974 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit E |
Gene Aliases: |
Atp6k, Atp6v0e1, M9.2 |
Location: |
Chr.17:26676369-26699647 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0e | NM_025272.2 | 1 | 90 | NP_079548.1 |
| AK007610.1 | 1 | 69 | BAB25133.1 | |
| AK008464.1 | 1 | 71 | ||
| AK013229.1 | 1 | 51 | BAB28727.1 | |
| AK017760.1 | 1 | 92 | BAB30911.1 | |
| AK030335.1 | 1 | 42 | BAC26909.1 | |
| AK134836.1 | 1 | 74 | BAE22305.1 | |
| AK159669.1 | 1 | 78 | BAE35273.1 | |
| AK168015.1 | 1 | 59 | BAE40002.1 | |
| BC024733.1 | 1 | 54 | AAH24733.1 | |
| BY707658.1 | 1 | 66 |
More Information
Set Membership: |
|

Genomic Map