Assay Details
Target Gene Details
Entrez Gene ID: | 75060 |
Gene Name: | RIKEN cDNA 4930506C21 gene |
Gene Aliases: |
- |
Location: |
Chr.17:8293366-8311118 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930506C21Rik | NR_073374.1 | |||
| AK015714.1 |
Target Gene Details
Entrez Gene ID: | 55951 |
Gene Name: | mitochondrial pyruvate carrier 1 |
Gene Aliases: |
0610006G08Rik, 3830411I18Rik, Brp44l |
Location: |
Chr.17:8283813-8297667 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpc1 | NM_018819.4 | NP_061289.1 | ||
| AF181116.1 | AAF25816.1 | |||
| AK002241.1 | BAB21958.1 | |||
| AK002266.1 | BAB21976.1 | |||
| AK004013.1 | BAB23124.1 | |||
| AK005348.1 | ||||
| AK013391.1 | BAB28826.1 | |||
| AK014421.1 | BAB29340.1 | |||
| AK014435.1 | BAB29347.1 | |||
| AK135943.1 | BAE22735.1 | |||
| AK170514.1 | BAE41851.1 | |||
| BC024365.1 | AAH24365.1 | |||
| BC103767.1 | AAI03768.1 |
More Information
Set Membership: |
|

Genomic Map