Assay Details
Target Gene Details
Entrez Gene ID: | 207932 |
Gene Name: | URB1 ribosome biogenesis 1 homolog (S. cerevisiae) |
Gene Aliases: |
4921511H13Rik, 5730405K23Rik, AA670781, AI645590, mKIAA0539 |
Location: |
Chr.16:90751527-90810465 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Urb1 | NM_029497.1 | NP_083773.1 | ||
| AK029516.1 | ||||
| AK220219.1 | BAD90144.1 |

Genomic Map