Assay Details
Target Gene Details
Entrez Gene ID: | 224833 |
Gene Name: | expressed sequence AI661453 |
Gene Aliases: |
- |
Location: |
Chr.17:47436639-47470638 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AI661453 | NM_145489.2 | NP_663464.2 | ||
| BC006605.1 | AAH06605.1 |

Genomic Map