Assay Details
Target Gene Details
Entrez Gene ID: | 320209 |
Gene Name: | DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11 |
Gene Aliases: |
4732462I11Rik, CHL1, CHLR1, KRG2, essa15a |
Location: |
Chr.17:66123501-66152174 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 25 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx11 | NM_001003919.1 | NP_001003919.1 | ||
| AK049419.1 | ||||
| AK076357.1 | ||||
| AK152868.1 | ||||
| BC049929.1 | AAH49929.1 | |||
| BC079656.1 | AAH79656.1 |
More Information
Set Membership: |
|

Genomic Map